Stampa…

o-Tolylacetic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Tolylacetic acid, (2-Methylphenyl)acetic acid

T0881-25G T0881-250G
TCIAT0881-25GEA 17 EUR
TCIAT0881-25G TCIAT0881-250G
o-Tolylacetic acid ≥98.0% (by titrimetric analysis)
o-Tolylacetic acid
Formula: (C₆H₄CH₃)CH₂COOH
Peso molecolare: 150,18 g/mol
Punto di ebollizione: 283 °C (1013 hPa)
Punto di fusione: 86…87 °C
MDL Number: MFCD00004328
CAS Number: 644-36-0
EINECS: 211-416-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=484625adc69222a03d47a7bc909c15ac28171faa&vid=484625adc69222a03d47a7bc909c15ac28171faa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 87.0 to 90.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR