(±)-1-Phenyl-2,2,2-trifluoroethanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
α-(Trifluoromethyl)benzyl alcohol, 2,2,2-Trifluoro-1-phenylethan-1-ol
T1792-25G
T1792-5G
TCIAT1792-25GEA
322
EUR
TCIAT1792-25G
TCIAT1792-5G
(±)-1-Phenyl-2,2,2-trifluoroethanol ≥98.0% (by GC)
(±)-1-Phenyl-2,2,2-trifluoroethanol
Formula:
C₆H₅CH(OH)CF₃ Peso molecolare: 176,14 g/mol Punto di ebollizione: 88°C Flash Pt: 84°C |
MDL Number:
MFCD00004498 CAS Number: 340-05-6 EINECS: 206-429-1 |
Risultati del test di specificazione
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...