(1S)-(-)-Borneol ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(-)-Borneol, L(-)-Borneol, (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
B1012-25G
B1012-500G
TCIAB1012-25GEA
25
EUR
TCIAB1012-25G
TCIAB1012-500G
(1S)-(-)-Borneol ≥95.0% (by GC)
(1S)-(-)-Borneol
Formula:
C₁₀H₁₈O Peso molecolare: 154,25 g/mol Punto di ebollizione: 203 °C (1013 hPa) Densità: 1,011 g/cm³ (20 °C) |
MDL Number:
MFCD00003759 CAS Number: 464-45-9 EINECS: 207-353-1 UN: 1312 ADR: 4.1,III Merck Index: 14,01338 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 95.0 % |
Specific rotation [a]20/D | -33.0 to -40.0 deg (C=5, EtOH) |
Solubility in EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...