D-(-)-N-Methylglucamine ≥99.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
Formula:
H₃CNHCH₂(CH(OH))₄CH₂OH Peso molecolare: 195.22 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004707 CAS Number: 6284-40-8 EINECS: 228-506-9 Merck Index: 14,06078 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 129.0 to 132.0 deg-C |
Specific rotation [a]20/D | -16.0 to -17.0 deg (C=2, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...