Stampa…

D-(+)-Homoserine ≥96.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-D-Hser-OH, H-D-HomoSer-OH, H-D-Homoser-OH, (R)-(+)-Homoserine, (R)-2-Amino-4-hydroxybutyric acid, D-Homoserine

H1224-1G H1224-5G
TCIAH1224-1GEA 56 EUR
TCIAH1224-1G TCIAH1224-5G
D-(+)-Homoserine ≥96.0% (by titrimetric analysis)
D-(+)-Homoserine
Formula: C₄H₉NO₃
Peso molecolare: 119,12 g/mol
Punto di fusione: 204°C
MDL Number: MFCD00077786
CAS Number: 6027-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d1eedd72c113177fb8edd18f27b64696915a27&vid=c8d1eedd72c113177fb8edd18f27b64696915a27&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D +6.0 to +10.0 deg (C=2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR