Stampa…

4-Bromo-α-methylbenzyl alcohol ≥99.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 4-Bromophenyl methyl carbinol, 1-(4-Bromophenyl)ethanol, p-Bromo-α-methylbenzyl alcohol

B1955-25G B1955-5G
TCIAB1955-25GEA 110 EUR
TCIAB1955-25G TCIAB1955-5G
4-Bromo-α-methylbenzyl alcohol ≥99.0% (by GC)
4-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
Punto di ebollizione: 134°C
Densità: 1,469 g/cm³ (15.2 °C)
Flash Pt: 63°C
MDL Number: MFCD00004514
CAS Number: 5391-88-8
EINECS: 226-389-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38992394dbef8a8959af0d969293e8c074bcbe4f&vid=38992394dbef8a8959af0d969293e8c074bcbe4f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR