Stampa…

(±)-3-Chloromandelic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: (±)-3-Chlorophenylglycolic acid, 3-Chloromandelic acid

C2009-25G C2009-5G
TCIAC2009-25GEA 110 EUR
TCIAC2009-25G TCIAC2009-5G
(±)-3-Chloromandelic acid ≥97.0% (by GC, titration analysis)
(±)-3-Chloromandelic acid
Formula: C₈H₇ClO₃
Peso molecolare: 186.59 g/mol
MDL Number: MFCD00829409
CAS Number: 16273-37-3
EINECS: 240-376-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e46a9bfbe0201a45c40b42001560607afa86bb59&vid=e46a9bfbe0201a45c40b42001560607afa86bb59&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -1.0 to +1.0 deg (C=1, MeOH)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR