(±)-2-Methylenanthic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(±)-2-Methylheptanoic acid, 2-Methylheptanoic acid
M1265-25G
M1265-5G
TCIAM1265-25GEA
83
EUR
TCIAM1265-25G
TCIAM1265-5G
(±)-2-Methylenanthic acid ≥98.0% (by GC, titration analysis)
(±)-2-Methylenanthic acid
Formula:
C₈H₁₆O₂ Peso molecolare: 144,21 g/mol Punto di ebollizione: 140°C Densità: 0,9122 g/cm³ (20 °C) |
MDL Number:
MFCD00142944 CAS Number: 1188-02-9 EINECS: 214-704-2 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...