Acido gibberellico ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
G0029-1G
G0029-100MG
G0029-5G
TCIAG0029-1GEA
30
EUR
TCIAG0029-1G
TCIAG0029-100MG
TCIAG0029-5G
Acido gibberellico ≥98.0% (by titrimetric analysis)
Acido gibberellico
Formula:
C₁₉H₂₂O₆ Peso molecolare: 346,38 g/mol Punto di fusione: 234 °C Densità: 1,386 g/cm³ (20 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 EINECS: 201-001-0 Merck Index: 14,04419 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +76.0 to +86.0 deg (C=2, Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...