Stampa…

1,3,5-Triazine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: s-Triazine

T0339-1G T0339-25G T0339-5G
TCIAT0339-1GEA 41 EUR
TCIAT0339-1G TCIAT0339-25G TCIAT0339-5G
1,3,5-Triazine ≥98.0% (by GC)
1,3,5-Triazine
Formula: C₃H₃N₃
Peso molecolare: 81,08 g/mol
Punto di ebollizione: 114°C
MDL Number: MFCD00006044
CAS Number: 290-87-9
EINECS: 206-028-1
ADR: 6.1,II
Merck Index: 14,09602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2c1464500f80e44f26e9873ee6931d8f08654db&vid=a2c1464500f80e44f26e9873ee6931d8f08654db&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 95.0 %
Melting point 79.0 to 83.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR