Stampa…

2-Hydroxy-3-nitropyridine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Nitropyridin-2(1H)-one, 3-Nitro-2-pyridinol

H0999-25G H0999-5G
TCIAH0999-25GEA 110 EUR
TCIAH0999-25G TCIAH0999-5G
2-Hydroxy-3-nitropyridine ≥98.0% (by HPLC, titration analysis)
2-Hydroxy-3-nitropyridine
Formula: C₅H₄N₂O₃
Peso molecolare: 140,1 g/mol
Punto di fusione: 224 °C
MDL Number: MFCD00006270
CAS Number: 6332-56-5
EINECS: 228-709-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e76c975e149a673cf8803491ebbc04081bd4861&vid=9e76c975e149a673cf8803491ebbc04081bd4861&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 224.0 to 228.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR