2-(Methylthio)pyrazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methylthiopyrazine
M1821-25G
M1821-5G
TCIAM1821-25GEA
41
EUR
TCIAM1821-25G
TCIAM1821-5G
2-(Methylthio)pyrazine ≥98.0% (by GC)
2-(Methylthio)pyrazine
Formula:
C₅H₆N₂S Peso molecolare: 126,18 g/mol |
MDL Number:
MFCD00041024 CAS Number: 21948-70-9 EINECS: 244-675-1 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...