1-Amino-2-methylnaphthalene ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methylnaphthalen-1-amine, 2-Methyl-1-naphthylamine
A0917-25G
A0917-5G
TCIAA0917-25GEA
217
EUR
TCIAA0917-25G
TCIAA0917-5G
1-Amino-2-methylnaphthalene ≥98.0% (by GC, titration analysis)
1-Amino-2-methylnaphthalene
Formula:
C₁₁H₁₁N Peso molecolare: 157,22 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00059135 CAS Number: 2246-44-8 |
Risultati del test di specificazione
Appearance | White or Colorless to Brown powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...