4-Bromo-1-naphthylamine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Amino-4-bromonaphthalene
A1886-25G
A1886-5G
TCIAA1886-25GEA
106
EUR
TCIAA1886-25G
TCIAA1886-5G
4-Bromo-1-naphthylamine ≥98.0% (by GC)
4-Bromo-1-naphthylamine
Formula:
C₁₀H₈BrN Peso molecolare: 222,08 g/mol |
MDL Number:
MFCD00004023 CAS Number: 2298-07-9 EINECS: 218-944-9 |
Risultati del test di specificazione
Appearance | White to Amber to Dark purple powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...