Triphenylphosphine oxide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
T0625-25G
T0625-500G
T0625-100G
TCIAT0625-25GEA
18
EUR
TCIAT0625-25G
TCIAT0625-500G
TCIAT0625-100G
Triphenylphosphine oxide ≥98.0% (by GC)
Triphenylphosphine oxide
Formula:
C₁₈H₁₅OP Peso molecolare: 278,29 g/mol Punto di ebollizione: ? 360 °C (1013 hPa) Punto di fusione: 155…158 °C Densità: 1,2 g/cm³ (20 °C) Flash Pt: 180 °C |
MDL Number:
MFCD00002080 CAS Number: 791-28-6 EINECS: 212-338-8 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...