Stampa…

Tetrapentylammonium iodide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetra-n-pentylammonium iodide, Tetraamylammonium iodide

T1011-25G T1011-5G
TCIAT1011-25GEA 140 EUR
TCIAT1011-25G TCIAT1011-5G
Tetrapentylammonium iodide ≥98.0% (by titrimetric analysis)
Tetrapentylammonium iodide
Formula: C₂₀H₄₄IN
Peso molecolare: 425,48 g/mol
Punto di fusione: 135…137 °C
Densità: 1,18 g/cm³ (25 °C)
MDL Number: MFCD00041980
CAS Number: 2498-20-6
EINECS: 219-688-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=245150758981c440837f15ac69ecbdfea3e4ff24&vid=245150758981c440837f15ac69ecbdfea3e4ff24&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 135.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR