Stampa…

Tetrahexilammonio ioduro ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetra-n-hexylammonium iodide

T1010-25G T1010-5G
TCIAT1010-25GEA 97 EUR
TCIAT1010-25G TCIAT1010-5G
Tetrahexilammonio ioduro ≥98.0% (by titrimetric analysis)
Tetrahexilammonio ioduro
Formula: C₂₄H₅₂IN
Peso molecolare: 481,59 g/mol
Punto di fusione: 105,4 °C
Densità: 1,0922 g/cm³ (25 °C)
MDL Number: MFCD00041981
CAS Number: 2138-24-1
EINECS: 218-382-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa77a6629c2b271b93af87da0eb8461431b8d572&vid=aa77a6629c2b271b93af87da0eb8461431b8d572&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 102.0 to 106.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR