Tetraethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Tetraethylammonium borohydride, Tetraethylammonium tetrahydridoborate
T0837-25G
T0837-5G
TCIAT0837-25GEA
181
EUR
TCIAT0837-25G
TCIAT0837-5G
Tetraethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)
Tetraethylammonium tetrahydroborate
Formula:
C₈H₂₄BN Peso molecolare: 145,1 g/mol |
MDL Number:
MFCD00031600 CAS Number: 17083-85-1 EINECS: 241-140-4 UN: 3131 ADR: 4.3,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Sodium hypochlorite Method) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...