2',3',4'-Trihydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(2,3,4-Trihydroxyphenyl)ethanone, Gallacetophenone
T1198-25G
T1198-5G
TCIAT1198-25GEA
74
EUR
TCIAT1198-25G
TCIAT1198-5G
2',3',4'-Trihydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
2',3',4'-Trihydroxyacetophenone
Formula:
C₈H₈O₄ Peso molecolare: 168,15 g/mol Punto di fusione: 173 °C |
MDL Number:
MFCD00002193 CAS Number: 528-21-2 EINECS: 208-430-2 Merck Index: 14,04342 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...