2',6'-Dichloro-3'-fluoroacetophenone ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D2750-25G
D2750-5G
TCIAD2750-25GEA
66
EUR
TCIAD2750-25G
TCIAD2750-5G
2',6'-Dichloro-3'-fluoroacetophenone ≥97.0% (by GC)
2',6'-Dichloro-3'-fluoroacetophenone
Formula:
C₈H₅Cl₂FO Peso molecolare: 207,03 g/mol |
MDL Number:
MFCD02093760 CAS Number: 290835-85-7 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...