Stampa…

ω-Bromo-4-nitroacetophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone

N0473-25G N0473-5G N0473-100G
TCIAN0473-25GEA 36 EUR
TCIAN0473-25G TCIAN0473-5G TCIAN0473-100G
ω-Bromo-4-nitroacetophenone ≥98.0% (by GC)
ω-Bromo-4-nitroacetophenone
Formula: C₈H₆BrNO₃
Peso molecolare: 244,04 g/mol
Punto di fusione: 100 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007356
CAS Number: 99-81-0
EINECS: 202-789-9
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c88f5b53c4ca0399c4faeb9186f22334d645a47&vid=2c88f5b53c4ca0399c4faeb9186f22334d645a47&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR