Stampa…

ω-Bromo-3-nitroacetophenone ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone

B2855-25G B2855-5G
TCIAB2855-25GEA 88 EUR
TCIAB2855-25G TCIAB2855-5G
ω-Bromo-3-nitroacetophenone ≥97.0% (by GC, titration analysis)
ω-Bromo-3-nitroacetophenone
Formula: C₈H₆BrNO₃
Peso molecolare: 244,04 g/mol
Punto di fusione: 90…94 °C
MDL Number: MFCD00024512
CAS Number: 2227-64-7
EINECS: 218-764-0
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 92.0 to 96.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR