Stampa…

Benzenesulphonamide ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

B0028-25G B0028-500G
TCIAB0028-25GEA 30 EUR
TCIAB0028-25G TCIAB0028-500G
Benzenesulphonamide ≥98.0% (by total nitrogen basis)
Benzenesulphonamide
Formula: C₆H₇NO₂S
Peso molecolare: 157,19 g/mol
Punto di fusione: 149…152 °C
MDL Number: MFCD00007930
CAS Number: 98-10-2
EINECS: 202-637-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ccd4216e30fd98811217ce8a54b1a2a0d1842c7&vid=7ccd4216e30fd98811217ce8a54b1a2a0d1842c7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 151.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR