Stampa…

(Methylsulphonyl)benzene ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl phenyl sulphone

M0401-25G M0401-500G
TCIAM0401-25GEA 71 EUR
TCIAM0401-25G TCIAM0401-500G
(Methylsulphonyl)benzene ≥97.0% (by GC)
(Methylsulphonyl)benzene
Formula: C₇H₈O₂S
Peso molecolare: 156,21 g/mol
MDL Number: MFCD00014741
CAS Number: 3112-85-4
EINECS: 221-476-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c268e10ab2d0b528d3d7722cbd504ef33f5e8fa&vid=0c268e10ab2d0b528d3d7722cbd504ef33f5e8fa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 86.0 to 89.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR