4-Fluoro-2-methoxyphenol ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₇H₇FO₂ Flash Pt: 101°C |
MDL Number:
MFCD00070797 CAS Number: 450-93-1 |
|
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...