Stampa…

Dimethyl-5-hydroxyisophthalate ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 5-Hydroxyisophthalic acid dimethyl ester, Dimethyl 5-hydroxyisophthalate

H1007-25G H1007-250G
TCIAH1007-25GEA 31 EUR
TCIAH1007-25G TCIAH1007-250G
Dimethyl-5-hydroxyisophthalate ≥98.0% (by GC, titration analysis)
Dimethyl-5-hydroxyisophthalate
Formula: C₁₀H₁₀O₅
Peso molecolare: 210.19 g/mol
MDL Number: MFCD00134367
CAS Number: 13036-02-7
EINECS: 235-899-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21284d679915ebaf5f238ce1c80b9aebd8103c79&vid=21284d679915ebaf5f238ce1c80b9aebd8103c79&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR