Stampa…

Monomethyl-5-nitroisophthalate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 5-Nitroisophthalic acid monomethyl ester, Methyl 5-nitroisophthalate

N0641-25G N0641-500G
TCIAN0641-25GEA 23 EUR
TCIAN0641-25G TCIAN0641-500G
Monomethyl-5-nitroisophthalate ≥98.0% (by titrimetric analysis)
Monomethyl-5-nitroisophthalate
Formula: C₉H₇NO₆
Peso molecolare: 225,16 g/mol
Punto di fusione: 177…180 °C
MDL Number: MFCD00009793
CAS Number: 1955-46-0
EINECS: 217-793-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50fadb7b12ee304975e947bebb722c35a8079783&vid=50fadb7b12ee304975e947bebb722c35a8079783&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 181.0 to 185.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR