4-Bromo-2-nitrophenol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B2363-25G
B2363-5G
TCIAB2363-25GEA
126
EUR
TCIAB2363-25G
TCIAB2363-5G
4-Bromo-2-nitrophenol ≥98.0% (by GC, titration analysis)
4-Bromo-2-nitrophenol
Formula:
C₆H₄BrNO₃ Peso molecolare: 218,01 g/mol Punto di ebollizione: 259 °C (1013 hPa) Punto di fusione: 90…94 °C |
MDL Number:
MFCD00082540 CAS Number: 7693-52-9 EINECS: 231-707-4 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...