Stampa…

4-Bromoisophthalic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B1260-10G B1260-25G
TCIAB1260-10GEA 151 EUR
TCIAB1260-10G TCIAB1260-25G
4-Bromoisophthalic acid ≥97.0% (by GC, titration analysis)
4-Bromoisophthalic acid
Formula: C₈H₅BrO₄
Punto di fusione: 303…304 °C
MDL Number: MFCD00002404
CAS Number: 6939-93-1
EINECS: 230-078-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e09ae9075cb3e7ea03a52a77cd1b0a73e386c05&vid=1e09ae9075cb3e7ea03a52a77cd1b0a73e386c05&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR