Stampa…

Penicillina G sale sodico ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

P1770-1G P1770-5G
TCIAP1770-1GEA 32 EUR
TCIAP1770-1G TCIAP1770-5G
Penicillina G sale sodico ≥98.0% (by HPLC, total nitrogen)
Penicillin G sodium salt
Formula: C₁₆H₁₇N₂NaO₄S
Peso molecolare: 356,38 g/mol
Punto di fusione: 227°C
Densità: 1,38 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00069666
CAS Number: 69-57-8
EINECS: 200-710-2
Merck Index: 14,07094
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f06b22f64bd2326d8accef847c0f1e27b12aeecc&vid=f06b22f64bd2326d8accef847c0f1e27b12aeecc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +285 to +310 deg (C=2, H2O) (calcd.on dried substance)
Drying loss max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR