Stampa…

4-Phenylmorpholine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: N-Phenylmorpholine

P0196-25G P0196-100G
TCIAP0196-25GEA 29 EUR
TCIAP0196-25G TCIAP0196-100G
4-Phenylmorpholine ≥98.0% (by GC, titration analysis)
4-Phenylmorpholine
Formula: C₁₀H₁₃NO
Punto di ebollizione: 270°C
Punto di fusione: 51…54 °C
Densità: 1,058 g/cm³ (20 °C)
MDL Number: MFCD00006166
CAS Number: 92-53-5
EINECS: 202-164-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8444787caf54ecb9bfde0533877c62bfcf4e0a4&vid=d8444787caf54ecb9bfde0533877c62bfcf4e0a4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR