α-Linolenic acid methyl ester ≥70% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl cis,cis,cis-9,12,15-octadecatrienoate, Methyl linolenate, 9Z,12Z,15Z-Octadecatrienoic acid methyl ester, Methyl Linolenate, Linolenic acid methyl ester, cis,cis,cis-9,12,15-Octadecatrienoic acid methyl ester, Methyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, Methyl α-linolenate
Formula:
C₁₉H₃₂O₂ Peso molecolare: 292,46 g/mol Punto di ebollizione: 347°C Punto di fusione: –57 °C Densità: 0,8982 g/cm³ (20 °C) |
MDL Number:
MFCD00135851 CAS Number: 301-00-8 EINECS: 206-102-3 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 70.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...