Benzyl butyrate ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Butyric acid benzyl ester, Butanoic acid benzyl ester
B0756-25G
B0756-500G
TCIAB0756-25GEA
16
EUR
TCIAB0756-25G
TCIAB0756-500G
Benzyl butyrate ≥97.0% (by GC)
Benzyl butyrate
Formula:
C₁₁H₁₄O₂ Peso molecolare: 178,23 g/mol Punto di ebollizione: 240°C Densità: 1,01 g/cm³ (20 °C) Flash Pt: 114°C |
MDL Number:
MFCD00027133 CAS Number: 103-37-7 EINECS: 203-105-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...