Benzyl acetoacetate ≥95.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₁H₁₂O₃ Punto di ebollizione: 163°C Densità: 1,11 g/cm³ (20 °C) Flash Pt: 108°C |
MDL Number:
MFCD00008785 CAS Number: 5396-89-4 EINECS: 226-416-4 |
|
Risultati del test di specificazione
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...