Ethyl isobutyrate ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Methylpropionic acid ethyl ester
I0105-25ML
I0105-500ML
TCIAI0105-25MLEA
16
EUR
TCIAI0105-25ML
TCIAI0105-500ML
Ethyl isobutyrate ≥99.0% (by GC)
Ethyl isobutyrate
Formula:
C₆H₁₂O₂ Peso molecolare: 116,16 g/mol Punto di ebollizione: 110°C Punto di fusione: –88 °C Densità: 0,87 g/cm³ (20 °C) Flash Pt: 15°C |
MDL Number:
MFCD00009165 CAS Number: 97-62-1 EINECS: 202-595-4 UN: 2385 ADR: 3,II Merck Index: 14,03814 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...