Ethyl 2-bromoisobutyrate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Ethyl α-bromoisobutyrate, Ethyl 2-bromo-2-methylpropionate, 2-Brom-2-methylpropionic acid ethyl ester
B0606-25G
B0606-500G
B0606-100G
TCIAB0606-25GEA
17
EUR
TCIAB0606-25G
TCIAB0606-500G
TCIAB0606-100G
Ethyl 2-bromoisobutyrate ≥98.0% (by GC)
Ethyl 2-bromoisobutyrate
Formula:
C₆H₁₁BrO₂ Peso molecolare: 195,06 g/mol Punto di ebollizione: 161°C Densità: 1,31 g/cm³ (20 °C) Flash Pt: 60°C |
MDL Number:
MFCD00000123 CAS Number: 600-00-0 EINECS: 209-980-6 UN: 2920 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...