Stampa…

Ethyl tridecanoate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Tridecylic acid ethyl ester

T0959-25ML T0959-5ML
TCIAT0959-25MLEA 248 EUR
TCIAT0959-25ML TCIAT0959-5ML
Ethyl tridecanoate ≥97.0% (by GC)
Ethyl tridecanoate
Formula: C₁₅H₃₀O₂
Peso molecolare: 242,4 g/mol
Punto di ebollizione: 198°C
Densità: 0,86 g/cm³ (20 °C)
MDL Number: MFCD00008978
CAS Number: 28267-29-0
EINECS: 248-929-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e7fcc22a2551322d055387c85c148afb8c8f864&vid=1e7fcc22a2551322d055387c85c148afb8c8f864&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR