Ethyl palmitate ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester
P0003-25ML
P0003-500ML
TCIAP0003-25MLEA
0
EUR
TCIAP0003-25ML
TCIAP0003-500ML
Ethyl palmitate ≥97.0% (by GC)
Ethyl palmitate
Formula:
C₁₈H₃₆O₂ Peso molecolare: 284,48 g/mol Punto di ebollizione: 342 °C (1013 hPa) Punto di fusione: 24 °C Densità: 0,86 g/cm³ (20 °C) |
MDL Number:
MFCD00008996 CAS Number: 628-97-7 EINECS: 211-064-6 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...