Ethyl decanoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester
D0022-25ML
D0022-500ML
TCIAD0022-25MLEA
16
EUR
TCIAD0022-25ML
TCIAD0022-500ML
Ethyl decanoate ≥98.0% (by GC)
Ethyl decanoate
Formula:
C₁₂H₂₄O₂ Peso molecolare: 200,32 g/mol Punto di ebollizione: 243°C Punto di fusione: –20 °C Densità: 0,862 g/cm³ (20 °C) Flash Pt: 123°C |
MDL Number:
MFCD00009581 CAS Number: 110-38-3 EINECS: 203-761-9 Merck Index: 14,03776 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...