Allyl enantate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Heptylic acid allyl ester, Enanthic acid allyl ester, Allyl Heptanoate, Allyl heptanoate, Heptanoic acid allyl ester
A2119-25G
A2119-250G
TCIAA2119-25GEA
79
EUR
TCIAA2119-25G
TCIAA2119-250G
Allyl enantate ≥98.0% (by GC)
Allyl enantate
Formula:
C₁₀H₁₈O₂ Peso molecolare: 170,25 g/mol Punto di ebollizione: 89°C Punto di fusione: –66°C Densità: 0,88 g/cm³ (20 °C) Flash Pt: 82°C |
MDL Number:
MFCD00038341 CAS Number: 142-19-8 EINECS: 205-527-1 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...