Stampa…

Heptanoyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

H0040-25G H0040-500G H0040-100G
TCIAH0040-25GEA 25 EUR
TCIAH0040-25G TCIAH0040-500G TCIAH0040-100G
Heptanoyl chloride ≥98.0% (by GC, titration analysis)
Heptanoyl chloride
Formula: C₇H₁₃ClO
Punto di ebollizione: 175°C
Densità: 0,9669 g/cm³ (15 °C)
Flash Pt: 58°C
MDL Number: MFCD00000763
CAS Number: 2528-61-2
EINECS: 219-775-3
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35c11222465d50041f7b06af58454ecdbc0ee470&vid=35c11222465d50041f7b06af58454ecdbc0ee470&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR