Isopropyl acetoacetate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Isopropyl 3-oxobutanoate
I0826-25G
I0826-500G
TCIAI0826-25GEA
32
EUR
TCIAI0826-25G
TCIAI0826-500G
Isopropyl acetoacetate ≥97.0% (by GC)
Isopropyl acetoacetate
Formula:
C₇H₁₂O₃ Peso molecolare: 144,17 g/mol Punto di ebollizione: 187°C Flash Pt: 71°C |
MDL Number:
MFCD00043933 CAS Number: 542-08-5 EINECS: 208-798-4 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...