Stampa…
Methyl trichloroacetate ≥96.0% (by GC)
Fornitore: TCI
Pericolo
Synonyms: Trichloroacetic acid methyl ester, TCA-methyl ester
T1013-25G T1013-500G
TCIAT1013-25GEA 41 EUR
TCIAT1013-25G TCIAT1013-500G
Methyl trichloroacetate ≥96.0% (by GC)
Methyl trichloroacetate
Formula: C₃H₃Cl₃O₂
Punto di ebollizione: 154°C
Punto di fusione: –17,5 °C
Densità: 1,4897 g/cm³ (20 °C)
Flash Pt: 72°C
MDL Number: MFCD00000794
CAS Number: 598-99-2
EINECS: 209-960-7
UN: 2533
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feeb2bf6d5152ec1fc075fd6b6573dfb263e0ed0&vid=feeb2bf6d5152ec1fc075fd6b6573dfb263e0ed0&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %