Stampa…

Tribromoacetic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

T0351-25G T0351-500G
TCIAT0351-25GEA 49 EUR
TCIAT0351-25G TCIAT0351-500G
Tribromoacetic acid ≥98.0% (by titrimetric analysis)
Tribromoacetic acid
Formula: Br₃CCOOH
Punto di ebollizione: 245 °C (1013 hPa)
Punto di fusione: 135 °C
MDL Number: MFCD00004167
CAS Number: 75-96-7
EINECS: 200-919-9
UN: 3261
ADR: 8,II
Merck Index: 14,09609
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0b65700b879764f1bd9d7cb6cb5359d618efeeb&vid=d0b65700b879764f1bd9d7cb6cb5359d618efeeb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR