4-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Phenyl-3-thiosemicarbazide, N-Phenylhydrazinecarbothioamide, 4-Phenylthiosemicarbazide
P0235-1G
P0235-25G
TCIAP0235-1GEA
16
EUR
TCIAP0235-1G
TCIAP0235-25G
4-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)
4-Phenyl-3-thiosemicarbazide
Formula:
C₇H₉N₃S Peso molecolare: 167,23 g/mol Punto di ebollizione: 308 °C (1013 hPa) Punto di fusione: 138…141 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007615 CAS Number: 5351-69-9 EINECS: 226-329-1 UN: 2811 ADR: 6.1,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...