Stampa…

2-Bromo-5-phenylthiophene ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B3663-1G B3663-5G
TCIAB3663-1GEA 72 EUR
TCIAB3663-1G TCIAB3663-5G
2-Bromo-5-phenylthiophene ≥97.0% (by GC)
2-Bromo-5-phenylthiophene
Formula: C₁₀H₇BrS
Peso molecolare: 239,14 g/mol
MDL Number: MFCD00159575
CAS Number: 29488-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10521889bd8767eb9b0d3e35b60b76a98f09fe61&vid=10521889bd8767eb9b0d3e35b60b76a98f09fe61&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR