Stampa…

1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Methylbis(trimethylsilyloxy)silane

H1267-25ML H1267-250ML
TCIAH1267-25MLEA 35 EUR
TCIAH1267-25ML TCIAH1267-250ML
1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥98.0% (by GC)
1,1,1,3,5,5,5-Heptamethyltrisiloxane
Formula: C₇H₂₂O₂Si₃
Peso molecolare: 222,51 g/mol
Punto di ebollizione: 142°C
Flash Pt: 22°C
MDL Number: MFCD00048000
CAS Number: 1873-88-7
EINECS: 217-496-1
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e214975f2b7a2adb3239a3fc10b1df309dfb9fe&vid=1e214975f2b7a2adb3239a3fc10b1df309dfb9fe&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR