Stampa…

6-Hydroxynicotinic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

H0633-25G H0633-250G
TCIAH0633-25GEA 321 EUR
TCIAH0633-25G TCIAH0633-250G
6-Hydroxynicotinic acid ≥97.0% (by GC, titration analysis)
6-Hydroxynicotinic acid
Formula: C₆H₅NO₃
Peso molecolare: 139,11 g/mol
Punto di fusione: 304…306 °C
MDL Number: MFCD00006277
CAS Number: 5006-66-6
EINECS: 225-682-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a1c955dfa3fb7b4e8105190aec1fe7d402ec4b1&vid=5a1c955dfa3fb7b4e8105190aec1fe7d402ec4b1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Solubility in 1mol/L NaOH very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR