2-Aminonicotinic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid
A0994-25G
A0994-5G
TCIAA0994-25GEA
79
EUR
TCIAA0994-25G
TCIAA0994-5G
2-Aminonicotinic acid ≥98.0% (by titrimetric analysis)
2-Aminonicotinic acid
Formula:
C₆H₆N₂O₂ Peso molecolare: 138,13 g/mol Punto di fusione: 297 °C Densità: 1,545 g/cm³ (20 °C) |
MDL Number:
MFCD00006318 CAS Number: 5345-47-1 EINECS: 226-296-3 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Sodium hydroxide solution | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...