Stampa…

2,6-Dichloronicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

D3412-25G D3412-5G
TCIAD3412-25GEA 187 EUR
TCIAD3412-25G TCIAD3412-5G
2,6-Dichloronicotinic acid ≥98.0% (by GC, titration analysis)
2,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
Peso molecolare: 192 g/mol
MDL Number: MFCD00075583
CAS Number: 38496-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc1e3b879bb97c44e966ab7ef89c495cda3511fe&vid=dc1e3b879bb97c44e966ab7ef89c495cda3511fe&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR